Learn More
Epredia™ Signature Series™ Pen-Fix™
Product Code. 13338656 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 1 gal.
Unit Size:
Each
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13338656 5 gal. Each
10148438 4 x 1 gal. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13338656 Supplier Epredia™ Supplier No. 6105
  • ONLINE EXCLUSIVE PRICE
    207.00€
    202.00€ / Pack of 1
    Save 5.00€
    2% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Achieve fixation and dehydration of specimens simultaneously with Epredia™ Signature Series™ Pen-Fix™, a uniquely buffered alcoholic formalin fixative.
  • Excellent frozen section fixative
  • Performs well with fatty or thick tissue sections
  • Penetration rate three times faster than 10% Neutral Buffered Formalin
  • Can be used on all open or enclosed tissue processors and in the grossing room
  • Recommended for use in the second and/or third stations of the processor as a secondary fixative
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 gal.