Learn More
Epredia™ Concentrated Neutral Buffered Formalin
Product Code. 10260219 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
Unit Size:
1 pack
Product Code. 10260219
Brand: Epredia™ 5700
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use Epredia™ Concentrated Neutral Buffered Formalin for Histopathology fixation.
  • Concentrate reduces storage space and contact with formaldehyde
  • One gallon of concentrate to four gallons of distilled or deionized water ensures consistent results with those of the premixed 10% NBF
TRUSTED_SUSTAINABILITY
Specifications
Product Type Formalin
Quantity 1 gal.
For Use With (Application) Fixation
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Neutral Buffered Formalin (Concentrated)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H371-May cause damage to organs.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.