Learn More
Epredia™ Richard-Allan Scientific™ Differentiating Solution
Product Code. 12523627 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
Unit Size:
3.8 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12523627 1 gal. 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12523627 Supplier Epredia™ Supplier No. 74211
  • ONLINE EXCLUSIVE PRICE
    109.00€
    99.22€ / 3.8 L
    Save 9.78€
    8% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Obtain clear differentiation and contrast between the hematoxylin and eosin with Epredia™ Richard-Allan Scientific™ Differentiating Solution.
  • Reproducible batch results allow for consistent staining in regressive procedures
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hydrochloric Acid Solution
Quantity 1 gal.