Learn More
Epredia™ Parapel™ Paraffin Repellent
Product Code. 10444910 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 oz. pump bottle; 6/CS
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10444910 4 oz. pump bottle; 6/CS Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10444910 Supplier Epredia™ Supplier No. 2300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Protect against excess buildup of wax on countertops and instrumentation with Epredia™ Parapel™ Paraffin Repellent.
  • Low odor
  • Reduces daily cleaning times
  • Will not leave behind oily residue
  • Will not remove finish from surfaces
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4 oz. pump bottle; 6/CS