missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riboflavin-d7 (6-d1; 7,8-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Riboflavin-D7 (6-D,7,8-dimethyl-D6) |
| Molecular Formula | C17 D7 H13 N4 O6 |
| Quantity | 0.005 g |
| Synonym | Riboflavin-d7 (6-d1, 7,8-dimethyl-d6), Riboflavin-D7 (6-D,7,8-dimethyl-D6), Riboflavin-d7, Vitamin B2-d7, 1-Deoxy-1-[3,4-dihydro-7,8-di(methyl-d3)-2,4-dioxobenzo[g]pteridin-10(2 H)-yl-6-d]-D-ribitol |
| InChI Formula | InChI=1 S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25 H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1/i1D3,2D3,3 D |
| SMILES | [2 H]c1c2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc(c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 6-deuterio-10-[(2 S,3 S,4 R)-2,3,4,5-tetrahydroxypentyl]-7,8-bis(trideuteriomethyl)benzo[g]pteridine-2,4-dione |
| Molecular Weight (g/mol) | 383.407 |
| Formula Weight | 383.1822 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction