Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Product Code. 10290285 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Rheodyne Injector 7030
Rheodyne Injector 8125
Rheodyne Injector 9010
Rheodyne Injector 9125
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7000/7010/7040
Rheodyne Injectors 7000/7010/7040/7067
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7060/7066/9060
Rheodyne Injectors 7125/7126
Rheodyne Injectors 8125/8126
Material:
ETFE
PEEK
Vespel
Unit Size:
Each
Product Code. 10290285
Brand: Thermo Scientific™ 3725018
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Rotor Seals
Material PEEK
Description 3725-018
For Use With (Equipment) Rheodyne Injectors 3725/3725i/3725-038/3725i-038