Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Alle ansehen Thermo Scientific ProdukteBeschreibung
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
Spezifikation
Spezifikation
| Type | Replacement Rotor Seals |
| Material | ETFE |
| Description | 9125-082 |
| For Use With (Equipment) | Rheodyne Injector 9125 |