Learn More
Thermo Scientific™ Rheodyne™ Ports for Injector Models 7010 and 9010
Product Code. 10678445 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
PEEK Loop Filler Port
PEEK Needle Port
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10678445 PEEK Needle Port Each
10303075 PEEK Loop Filler Port Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10678445 Supplier Thermo Scientific™ Supplier No. 9013
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
Estimated Shipment: 02-12-2026
Add to basket
Edge
Add to basket
Use these ports for your Thermo Scientific™ Rheodyne™ 7010 and 9010 Sample Injectors for HPLC.
Use Thermo Scientific™ Rheodyne Ports for Models 7010 and 9010 Injectors for high-performance liquid chromatography (HPLC).
TRUSTED_SUSTAINABILITY
Specifications
Type PEEK Needle Port
For Use With (Application) Liquid Chromatography
For Use With (Equipment) Rheodyne HPLC Injector Models 7010 and 9010