Learn More
Thermo Scientific™ RheBuild™ Kit
Product Code. 11518550 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
7010 with Stator
Unit Size:
Each
Product Code. 11518550
Brand: Thermo Scientific™ 7010997
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Eliminate the need to order individual parts. Included in each individualized RheBuild Kit are all the parts, tools, and instructions to maintain the precision performance of Rheodyne stainless steel and PEEK platforms.
  • For Rheodyne™ 7010 with Stator
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 7010 with Stator
Model Rheodyne 7010
Type RheBuild Kit