Learn More
Reynosin, TRC
Product Code. 30012513
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012513 10 mg 10mg
30010501 2.5 mg 2.5mg
30003718 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012513 Supplier Toronto Research Chemicals Supplier No. TRCR67723010MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28254-53-7
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.32
Synonym (3aS,5aR,6R,9aS,9bS)-Decahydro-6-hydroxy-5a-methyl-3,9-bis(methylene)naphtho[1,2-b]furan-2(3H)-one1beta,6alpha-Dihydroxy-gamma-lactone-eudesma-4(14),11(13)-dien-12-oic AcidNSC 155623Reinosin
IUPAC Name (3aS,5aR,6R,9aS,9bS)-6-hydroxy-5a-methyl-3,9-dimethylidene-3a,4,5,6,7,8,9a,9b-octahydrobenzo[g][1]benzofuran-2-one
SMILES C[C@@]12CC[C@@H]3[C@H](OC(=O)C3=C)[C@H]1C(=C)CC[C@H]2O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C15H20O3/c1-8-4-5-11(16)15(3)7-6-10-9(2)14(17)18-13(10)12(8)15/h10-13,16H,1-2,4-7H2,3H3/t10-,11+,12+,13-,15-/m0/s1
Formula Weight 248.1412
Chemical Name or Material Reynosin