Learn More
Zeiss™ Revolver Reflector
Click to view available options
Magnification Power:
6X
Unit Size:
Each
Description
Compatible Microscopes:
- Axio Observer 3
- Axio Observer 3 materials
- Axio Observer 5
- Axio Observer.A1
- Axio Observer.A1 Entry
- Axio Observer.A1m
- Axio Observer.D1
- Axio Observer.D1 Entry
- Axio Observer.D1 Mid Range
- Axio Observer.D1m
- Axio Observer.Z1
- Axio Observer.Z1 High End
- PALM MicroTweezers Rel.4.2
Specifications
Specifications
| Type | 6-position Reflector Turret |
| For Use With (Equipment) | P&C modules |
| Magnification Power | 6X |
| Quantity | 1 Ea. |