Learn More
Zeiss™ Revolver Reflector
Product Code. 11815590
Click to view available options
Magnification Power:
6X
Unit Size:
Each
Product Code. 11815590
Brand: Zeiss™ 4249450000000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for P&C modules reflector modules. Modules exchangable without removal of reflector turret
Compatible Microscopes:
  • Axio Observer 3
  • Axio Observer 3 materials
  • Axio Observer 5
  • Axio Observer.A1
  • Axio Observer.A1 Entry
  • Axio Observer.A1m
  • Axio Observer.D1
  • Axio Observer.D1 Entry
  • Axio Observer.D1 Mid Range
  • Axio Observer.D1m
  • Axio Observer.Z1
  • Axio Observer.Z1 High End
  • PALM MicroTweezers Rel.4.2
Specifications
Type 6-position Reflector Turret
For Use With (Equipment) P&C modules
Magnification Power 6X
Quantity 1 Ea.