Learn More
RUBIS Reverse Action Tweezers
Product Code. p-9691002
Encompass_Preferred
Click to view available options
Tip Style:
Pointed, Ultra Fine
Round, Flat
Unit Size:
Each
Product Code. 11743426
Brand: RUBIS OR
Reverse Action Tweezers - Antimagnetic/Anti-acid SS
  • Reverse Action Tweezers
  • High Precision Tweezers
  • 100% Made in Switzerland
  • Push to open
  • Anti-acid/Antimagnetic Stainless Steel
  • Perfect alignment
  • Very Reliable
  • Designed for prolongated use applications
Specifications
Tip Style Pointed, Ultra Fine
Length (English) 4.52 in.
Material Anti-acid, Anti-magnetic Stainless Steel
Length (Metric) 115 mm
Product Type Reverse Action Tweezers
Quantity (No. per Case) Each
Type Auto-locking Tweezers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.