Learn More
Masterflex™ Reusable White Acetal Copolymer Hose Clamp
Product Code. 12409660
Click to view available options
For Use With:
0.23 x 0.26 in. Tubing I.D.
0.35 x 0.39 in. Tubing I.D.
0.40 x 0.45 in. Tubing I.D.
0.51 x 0.59 in. Tubing I.D.
0.61 x 0.70 in. Tubing I.D.
Unit Size:
Pack of 100
Product Code. 12409660
Brand: Masterflex™ 0683202
Specifications
Material Acetal Copolymer
For Use With 0.35 x 0.39 in. Tubing I.D.
Packaging Quantity 100 Pack
Color White
Product Type Hose Clamp