Learn More
3M™ Reusable Half Face Mask Respirator
Product Code. 17766983 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
Model:
HF-301
HF-302
HF-303
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Model Unit Size
17766983 Small HF-301 Pack of 10
17717003 Medium HF-302 Pack of 10
17746993 Large HF-303 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17766983 Supplier 3M™ Supplier No. 7100265024
  • ONLINE EXCLUSIVE PRICE
    579.00€
    398.53€ / Pack of 10
    Save 180.47€
    31% Off
Estimated Shipment: 20-01-2027
3M™ Reusable Half Mask has been designed with user comfort and convenience in mind. Compatible with DIN threaded, 3M™ Filters DT-Series, the HF-301 offers protection against a wide range of respiratory hazards.
  • Hybrid reflex seal contours and flexes to the user's face to aid wearer comfort
  • Seal check mechanism - when pressed during exhale, the mask inflates slightly showing that a good seal has been achieved
  • Integrated voice panel for enhanced voice intelligibility while wearing the mask
  • Nose pinch allows for improved integration with eyewear
  • Comfortable, durable, low profile head cradle & thick elasticated straps with simple rear fastening
TRUSTED_SUSTAINABILITY
Specifications
Product Type Half Facepiece Respirator
Size Small
For Use With (Equipment) DIN Threaded, 3M™ Filters DT-Series
Model HF-301
Quantity 10/Cs.