Learn More
Fisherbrand™ Reusable Class A Volumetric Pipets
Product Code. 17871440 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
2 mL
3 mL
4 mL
5 mL
Type:
Serialized Volumetric Pipet
Volumetric Pipet
Min. Flow Time:
8 sec.
Accuracy:
±0.006 mL
±0.01 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 17871440
Only null left
Add to basket
Add to basket
Available in either standard or serialized models.
  • Borosilicate glass
  • Volume accuracy marked on each pipet
  • Permanent amber markings
  • Tuf-Temp™–treated tempered tips reduce chipping
Order Info
All pipets are available as PersonaPet™ personalized pipets.
Specifications
Accuracy ±0.01 mL
Calibrated Yes
Certifications/Compliance ASTM E969-95
Class A
Material Borosilicate Glass
Graduated Yes
Graduation Color Amber
Min. Flow Time 8 sec.
Tip Style Tuf-Temp™ treated
Volume (English) 0.1 oz.
Color Black
Volume (Metric) 3 mL
Type Volumetric Pipet
Disposable Reusable