Learn More
RETSCH ZM200 Ultra Centrifugal Mill: Distance Sieves
Product Code. 10052212 Shop All Retsch Products
Click to view available options
Pore Size:
0.01 in. (0.50 mm)
0.08 mm
0.12 mm
0.20 mm
0.25 mm
0.75 mm
1.50 mm
4.0 mm
Pore Style:
Conidur holes
Round holes
Trapezoid holes
Unit Size:
Each
Product Code. 10052212
Brand: RETSCH 03.647.0260
Only null left
Add to basket
Add to basket
For temperature-sensitive materials
Specifications
Product Type Distance Sieve
For Use With (Equipment) ZM200 ultra centrifugal mil
Material Stainless Steel
Pore Size 1.50 mm
Pore Style Trapezoid holes