Learn More
RETSCH ZM 200 Mill Accessories
Product Code. 10370201 Shop All Retsch Products
Change view
Click to view available options
Product Type:
12-tooth Rotor
6-tooth Rotor
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10370201 6-tooth Rotor Each
10237940 12-tooth Rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10370201 Supplier RETSCH Supplier No. 02.608.0043
  • ONLINE EXCLUSIVE PRICE
    3725.00€
    3408.90€ / Pack of 1
    Save 316.10€
    8% Off
Estimated Shipment: 27-01-2027
Push-fit assembly, with wear-resistant coating
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
  • Stainless-steel, push-fit, with wear-resistant coating
  • 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
  • 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
Product Type 6-tooth Rotor
For Use With (Equipment) Ultra Centrifugal mill ZM 200
Includes Wear-resistant coating
Material Stainless Steel