Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Product Code. 10330622 Shop All Retsch Products
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
1 mm
106 μm
1.18 mm
125 μm
1.4 mm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
25 mm
Unit Size:
Each
Product Code. 10330622
Brand: RETSCH 60.150.001000
Stainless steel with wire gauze
Specifications
Depth (English) 2 in.
Depth (Metric) 5.08 cm
Pore Size 1 mm
Type Test Sieve
Certifications/Compliance ASTM E11
Diameter (English) Outer 8 in.
Material Stainless Steel
Tyler Standard 18 Mesh
U.S. Standard No. 18
For Use With (Application) Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
Diameter (Metric) Outer 8 in.
Mesh Material Stainless Steel