Learn More
RETSCH Comfort Clamping Device for AS 200 Sieve Shakers
Product Code. 10625441 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
AS 200 sieve shaker
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10625441 AS 200 sieve shaker Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10625441 Supplier RETSCH Supplier No. 32.662.0001
  • ONLINE EXCLUSIVE PRICE
    985.00€
    923.96€ / Pack of 1
    Save 61.04€
    6% Off
Estimated Shipment: 09-12-2026
Provides reliable, quick and convenient clamping of sieves in dry operation.
  • View cover, smooth rods, and quick-action clamps
  • Cover stays on rods
  • Removable sieve stack
Specifications
Holds Test Sieves, 200 to 203 mm dia.
Product Type Comfort Clamping Device
For Use With (Application) Reliable, quick, convenient sieve clamping in dry operation
For Use With (Equipment) AS 200 sieve shaker