Learn More
Miele Professional™ Membrane Pump and Suction Lance PLW8636 APLW 885
Shop All Miele Products
Click to view available options
Model:
APLW 885
Unit Size:
1 sheet
Description
- For 5 - 10 L canisters in the dispensing drawer
- The suction lance features level detection
- Installed by Customer Service
- Extra disp. canisters with installation by customer service
Specifications
Specifications
| Product Type | Dispensing System |
| Model | APLW 885 |
| For Use With (Equipment) | Washer-Disinfectors |
| Includes | 10 to 20 L Canister |
| Quantity | 1 Ea. |