Learn More
Brady™ Retractable Barrier Belt with Silver Posts
Product Code. 11761156 Shop All Brady Products
Change view
Click to view available options
Color:
Black/Yellow
Burgundy
Fluorescent Yellow
Green
Length (Metric):
2 m
3.5 m
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Unit Size
11761156 Black/Yellow 2 m Each
11751156 Burgundy 2 m Each
11711166 Green 3.5 m Each
11751166 Fluorescent Yellow 3.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11761156 Supplier Brady™ Supplier No. 912678
  • ONLINE EXCLUSIVE PRICE
    570.00€
    553.35€ / Pack of 1
    Save 16.65€
    2% Off
Estimated Shipment: 21-12-2026
Retractable Barrier Belt with Silver Posts for traffic control.
  • For traffic control.
Specifications
Product Type Stanchion
Alternate Part Number ADAPT FR BARRIER BELT 2M 804326 YL/BK
Color Black/Yellow
Material Steel
Description Retractable Barrier Belt - 2 Meters with Silver Posts
For Use With (Application) Traffic Control
Height (English) 3.94 in.
Height (Metric) 100 mm
Includes Retractable Barrier Belt (2 m), Silver Posts
Length (English) 6.6 ft.
Length (Metric) 2 m
Quantity 1 Ea.
Weight (Metric) 12 kg
Width (English) 3.94 in.
Width (Metric) 100 mm