Learn More
Brady™ Retractable Barrier Belt with Silver Posts
Product Code. 11711166
Click to view available options
Color:
Black/Yellow
Burgundy
Fluorescent Yellow
Green
Length (Metric):
2 m
3.5 m
Unit Size:
Each
Product Code. 11711166
Brand: Brady™ 912683
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Retractable Barrier Belt with Silver Posts for traffic control.
  • For traffic control.
Specifications
Product Type Stanchion
Color Green
Material Steel
Description Retractable Barrier Belt - 3.5 Meters with Silver Posts
For Use With (Application) Traffic Control
Height (English) 3.94 in.
Height (Metric) 100 mm
Includes Retractable Barrier Belt (3.5 m), Silver Posts
Length (English) 11.5 ft.
Length (Metric) 3.5 m
Quantity 1 Ea.
Weight (Metric) 12 kg
Width (English) 3.94 in.
Width (Metric) 100 mm