Learn More
Brady™ Retractable Barrier Belt with Chrome Posts
Product Code. 11791116 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Red
Length (Metric):
2 m
3.5 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Unit Size
11791116 Black 2 m Each
11781126 Red 3.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11791116 Supplier Brady™ Supplier No. 912641
  • ONLINE EXCLUSIVE PRICE
    581.00€
    553.35€ / Pack of 1
    Save 27.65€
    4% Off
Estimated Shipment: 27-10-2026
Retractable Barrier Belt with Chrome Posts for traffic control.
  • Traffic control barrier.
Specifications
Product Type Stanchion
Alternate Part Number ADAPT CHR BARRIER BELT 2M 804324 BLACK
Color Black
Material Steel
Description Retractable Barrier Belt - 2 Meters with Chrome Posts
For Use With (Application) Traffic Control
Height (English) 3.94 in.
Height (Metric) 100 mm
Includes Retractable Barrier Belt (2 m), Chrome Posts
Length (English) 6.6 ft.
Length (Metric) 2 m
Quantity 1 Ea.
Weight (Metric) 12 kg
Width (English) 3.94 in.
Width (Metric) 100 mm