Learn More
Brady™ Retractable Barrier Belt with Black Posts
Product Code. 11771146 Shop All Brady Products
Change view
Click to view available options
Color:
Burgundy
Fluorescent Yellow
Green
Length (Metric):
2 m
3.5 m
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Length (Metric) Unit Size
11771146 Burgundy 3.5 m Each
11771136 Green 2 m Each
11791146 Fluorescent Yellow 3.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11771146 Supplier Brady™ Supplier No. 912669
  • ONLINE EXCLUSIVE PRICE
    762.00€
    748.56€ / Pack of 1
    Save 13.44€
    1% Off
Estimated Shipment: 03-11-2026
Retractable Barrier Belt with Black Posts for traffic control.
  • For traffic control.
Tekniske data
Product Type Stanchion
Alternate Part Number ADAPT BK BARRIER BELT 3,7M 804329 DK RED
Color Burgundy
Material Steel
Description Retractable Barrier Belt - 3.5 Meters with Black Posts
For Use With (Application) Traffic Control
Height (English) 3.94 in.
Height (Metric) 100 mm
Includes Retractable Barrier Belt (3.5 m) Black Posts
Length (English) 11.5 ft.
Length (Metric) 3.5 m
Quantity 1 Ea.
Weight (Metric) 12 kg
Width (English) 3.94 in.
Width (Metric) 100 mm
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.