Learn More
Bochem™ Retort Clamps
Click to view available options
Material:
Laboral
Stainless Steel (18/10)
Jaw Type:
Red PVC-coated
Transparent Silicone-coated
Unit Size:
Each
Description
- Stainless Steel
- Adjustable
Specifications
Specifications
| Material | Stainless Steel (18/10) |
| Description | Span width (size) independently adjustable by 2 regulating screws |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 125 mm |
| Size (Metric) Grip | 8 to 30 mm |
| Jaw Type | Transparent Silicone-coated |