Learn More
Cytiva Research Grade FBS, Irradiated and Heat Inactivated
Product Code. 15745533
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15745533 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15745533 Supplier Cytiva Supplier No. SV30160.03IH
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Research grade Fetal Bovine Serum of South America Origin. Irradiated and Heat inactivated.
  • Supplement research cell culture growth with South America-sourced quality HyClone Fetal Bovine Serum.
  • Sourced in EU-approved South American countries with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100nm (0.1um) pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY
Specifications
Country of Origin South America
Quantity 500 mL
Grade Research grade