missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ReproRNA™-OKSGM

Product Code. 17151036
Encompass
Change view
Click to view available options
Quantity:
12 μg
Unit Size:
12µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17151036 12 μg 12µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17151036 Supplier STEMCELL Technologies Supplier No. 05931
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A non-integrating, self-replicating RNA reprogramming vector for generating iPS cells

  • ReproRNA™-OKSGM is a single-stranded RNA replicon vector that contains five reprogramming factors: OCT4, KLF-4, SOX2, GLIS1, and c-MYC, as well as a puromycin-resistance gene (Figure 1)
  • This RNA vector reprograms somatic cells, such as fibroblasts, into induced pluripotent stem (iPS) cells with high efficiency and only requires a single transfection step (Figure 2)
  • When used together with ReproTeSR™ reprogramming medium, the generation of iPS cell colonies can be achieved under feeder-free conditions with superior colony morphology and similar reprogramming efficiency to feeder-based systems (Figures 3 - 4)
  • ReproRNA™-derived iPS cell colonies also express markers of undifferentiated cells and retain a normal karyotype (Figures 5-6)
  • Subsequently, iPS cells generated with ReproRNA™-OKSGM can be maintained in TeSR™ maintenance media (mTeSR™1, TeSR™2, or TeSR™-E8™) and further differentiated into cells of all three germ layers (Figure 7)

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture; Reprogramming
Quantity 12 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.