Learn More
Sartorius™ ReproEasy Performance Test Pads
Product Code. 15689963 Shop All Sartorius Products
Click to view available options
Type:
Moisture Analyzer Test Pads, 10/Pk.
Unit Size:
Each
Pack of 10
Product Code. 15689963
Brand: Sartorius™ YHP01MA
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
  • Allows you to quickly confirm whether a device is working properly and delivers correct results.
  • Use together with the performance test function in moisture analyzer.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MA37/160 Moisture Analyzer
Quantity 10/Pk.
Type Moisture Analyzer Test Pads, 10/Pk.