Learn More
BRAND™ Replacement Wash Head
Click to view available options
Material: ETFE
Description
- Spare wash head
- GL 32
- With delivery tube
Specifications
Includes | FEP Delivery Tube |
Material | ETFE |
Type | Wash head |
Thread Size (Metric) | GL 32 |