Learn More
Trajan™ SilTite™ FingerTite Ferrule
For gas chromatography systems
Brand:  Trajan™ 073631
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15228983
  • 84.70€ / Pack of 10
Estimated Shipment: 28-06-2024
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
GC Ferrule
10/Pk.
0.5 mm