Learn More
Scienceware Mercury Collector Jar with Integrated Sponge and Perforated Plate
Product Code. 13064897 Shop All Cole-Parmer Products
Change view
Click to view available options
Type:
Mercury Spill Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13064897 Mercury Spill Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13064897 Supplier Scienceware Supplier No. 0670600
  • ONLINE EXCLUSIVE PRICE
    48.35€
    42.85€ / Pack of 1
    Save 5.50€
    11% Off
View all promotions
Estimated Shipment: 30-11-2026
The Mercury Collector Jar with Integrated Sponge and Perforated Plate makes small mercury spill clean-up quick and simple.
  • Foam pad on lid liner absorbs spilled mercury.
  • Simply screw on lid to collect mercury beneath perforated plate.
Specifications
Type Mercury Spill Kit
For Use With (Application) Mercury Collection
Length (English) 4 in.
Length (Metric) 10.16 cm
Width (English) 2 in.
Width (Metric) 5.08 cm