Learn More
DWK Life Sciences Replacement Seal
Product Code. 15837155 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Plastic Hose Connections (Cat. No. 29 25506 03 and 29 247 05 04)
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15837155 Plastic Hose Connections (Cat. No. 29 25506 03 and 29 247 05 04) Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15837155 Supplier DWK Life Sciences Supplier No. 292200904
  • ONLINE EXCLUSIVE PRICE
    8.58€
    7.80€ / Pack of 10
    Save 0.78€
    9% Off
Estimated Shipment: 13-10-2026
The sealing ring serves as an intermediate piece between the plastic olive and the glass thread and enables a leak-proof connection of the components (leakage protection).
A seal is part of a construction and prevents or limits unwanted mass transfer.
  • For leak-tight connection of the plastic olives
Specifications
For Use With (Equipment) Plastic Hose Connections (Cat. No. 29 25506 03 and 29 247 05 04)
Quantity 10/Pk.
Product Type Sealing Ring for Olive
Material Silicone (VMQ)