Learn More
Trajan Scientific and Medical™ Replacement Plungers for CTC MEPS Syringes
Replacement plungers for use with CTC and Thermo MEPS syringes.
Brand:  Trajan Scientific and Medical™ 0318301
Additional Details : Weight : 0.01000kg
Product Code. 15218553
  • 46.80€ / Each
Estimated Shipment: 10-07-2024
Description
  • Removable needle
  • With GT plunger
Specifications
Specifications
Replacement plunger
60 mm
250 μL
CTC and Thermo MEPS Syringes