Learn More
Merck Replacement Parts for Stirred Cells
Product Code. 15855568 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
Amicon 8003 Series Stirred Cell
Amicon 8200 Series Stirred Cell
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15855568 Amicon 8200 Series Stirred Cell Each
11700403 Amicon 8003 Series Stirred Cell 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15855568 Supplier Merck Supplier No. 8200SCPKIT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Suitable for use with Amicon™ stirred cell apparatus
  • Small Replacement Parts for 8000 Series Stirred Cells include 2 O-rings (Mfr. Nos. 2122S, 2118S, 2015S), tubing (Mfr. No. 4500202), tubing fitting assembly (Mfr. No. 15025AM, and membrane holder (Mfr. No. 2214801)
  • Large Replacement Parts for Stirred Cells include one each: cap assembly, stirrer assembly and base
TRUSTED_SUSTAINABILITY
Spezifikation
Includes Cap assembly (1524901), stirrer assembly (15247) and base (210000)
Type Replacement Part
For Use With (Equipment) Amicon 8200 Series Stirred Cell