Learn More
KD Scientific Replacement Parts for Stainless Steel Syringes
Product Code. 16174327
Click to view available options
Type:
1/16 in. Swagelok Fitting
O-ring
Tip Seal O-Ring
For Use With:
100 mL Syringe
20 mL Syringe
20 to 200 mL Syringe
20 to 200 mL Syringes
50 mL Syringe
Material:
Perfluoroelastomer
Stainless Steel
Viton
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Product Code. 16174327
Brand: KD Scientific 780843
Only null left
Add to basket
Add to basket
A well looked after stainless steel syringe can last a life time. KD Scientific provides replacement parts like O-rings, tip seals and various fittings to keep your syringes performing at the highest level.
  • Replacement seals
  • Perfluoroelastomer O-Rings
  • Perfluoroelastomer tip seal O-Rings
  • Viton O-Rings
  • Viton tip seal O-Rings
  • Swagelock™ fittings
  • Luer Lock Fittings
Specifications
For Use With 20 to 200 mL Syringe
IP Rating None
Material Viton
Type Tip Seal O-Ring