Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Product Code. 11772227 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo 365H2301/-F193/-G216/-H238/-I2611/-F1901/-G2091/-J2881 Syringes
Thermo 365K3041 Syringe
Thermo Scientific 36500490 Syringe
Thermo Scientific 36500495, 36503015 Syringes
Thermo Scientific 36500510 Syringe
Thermo Scientific 36500520 Syringe
Thermo Scientific 36550040 Syringe
Thermo Scientific 365D1171 Syringe
Thermo Scientific 365D1841 Syringe
Thermo Scientific 365D3731 Syringe
Length (Metric) Needle:
50 mm
75 mm
80 mm
Unit Size:
Pack of 2
Pack of 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Length (Metric) Needle Unit Size
11772227 Thermo Scientific 365D1171 Syringe 50 mm Pack of 5
10219214 Thermo Scientific 36550040 Syringe 50 mm Pack of 2
10260095 Thermo Scientific 36500510 Syringe 75 mm Pack of 2
10270095 Thermo Scientific 36500520 Syringe 75 mm Pack of 2
10371005 Thermo Scientific 36500490 Syringe 80 mm Pack of 5
10055864 Thermo Scientific 36500495, 36503015 Syringes 50 mm Pack of 5
10351405 Thermo 365H2301/-F193/-G216/-H238/-I2611/-F1901/-G2091/-J2881 Syringes 50 mm Pack of 5
10733287 Thermo 365K3041 Syringe 50 mm Pack of 5
10243033 Thermo Scientific 365D1841 Syringe 50 mm Pack of 2
10351245 Thermo Scientific 365D3731 Syringe 50 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 missing translation for 'options'
Product Code. 11772227 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 365RN215
Estimated Shipment: 07-12-2026
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Description 365RN215
Needle Gauge 26 G
Length (English) Needle 1.9685 in.
Product Type Replacement Syringe Needle
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
For Use With (Equipment) Thermo Scientific 365D1171 Syringe
Connection Type Removable