Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Product Code. 10260095 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo 365H2301/-F193/-G216/-H238/-I2611/-F1901/-G2091/-J2881 Syringes
Thermo 365K3041 Syringe
Thermo Scientific 36500490 Syringe
Thermo Scientific 36500495, 36503015 Syringes
Thermo Scientific 36500510 Syringe
Thermo Scientific 36500520 Syringe
Thermo Scientific 36550040 Syringe
Thermo Scientific 365D1171 Syringe
Thermo Scientific 365D1841 Syringe
Thermo Scientific 365D3731 Syringe
Length (Metric) Needle:
50 mm
75 mm
80 mm
Unit Size:
Pack of 2
Pack of 5
Product Code. 10260095
Brand: Thermo Scientific™ 36550045
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Description 36550045
Needle Gauge 31 G
Product Type Replacement Syringe Needle
Volume (Metric) 5 μL
Length (Metric) Needle 75 mm
Needle Point Style Cone
For Use With (Equipment) Thermo Scientific 36500510 Syringe
Connection Type Removable