Learn More
A.I.E Replacement Heating Element for Heat Sealers
Product Code. 13443719 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With (Application):
For Heat Sealer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
13443719 For Heat Sealer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13443719 Supplier A.I.E Supplier No. 0301885
  • ONLINE EXCLUSIVE PRICE
    61.15€
    51.15€ / Pack of 1
    Save 10.00€
    16% Off
Estimated Shipment: 22-01-2027
Quickly and easily replace the heating element of your heat sealer.
  • Model 03018-80 replacement heat element kit is compatible with 03018-10 heat sealer.
  • Model 03018-85 replacement heat element kit is compatible with 03018-15 heat sealer.
Specifications
Temperature (Metric) 399°C
Type Replacement Heating Element
For Use With (Application) For Heat Sealer
Length (Metric) 304.8 mm
Width (Metric) 5.08 mm
Voltage 230 V