Learn More
SCAT Replacement Dirt Sieve for Funnel MARCO V2.0
Shop All SCAT Europe Products
Click to view available options
For Use With (Equipment):
Funnel MARCO V2.0
Unit Size:
Each
Description
- Dirt sieve
- For funnel
- With lid MARCO, V2.0
- PE-HD
- Blue
Specifications
Specifications
| Material | HDPE-EC |
| Height (Metric) | 20 mm |
| Color | Black |
| For Use With (Equipment) | Funnel MARCO V2.0 |
| Diameter (English)Top | 140 mm |
| Product Type | Replacement Dirt Sieve |