Learn More
Electrothermal™ Replacement Digi-Mantle Heater Cartridge
Product Code. 11777873
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11777873 1000 mL Each
11757873 250 mL Each
11767873 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11777873 Supplier Electrothermal™ Supplier No. 3620409
  • ONLINE EXCLUSIVE PRICE
    1120.00€
    818.13€ / Pack of 1
    Save 301.87€
    26% Off
Estimated Shipment: 07-01-2027
Enables a round bottom flask to be stirred and heated up to an element temperature of 450°C. Electrothermal™ Replacement Digi-Mantle Heater Cartridge uses precise electronic control to provide accurate e of temperature and stirring measures.
  • For use with the OMCA Digital Controller.
  • Stir and heat round bottom flasks up to an element temperature of 450°C
  • Uses precise electronic control to give an accurate measure of temperature and stirring.
  • Modular design allows heating cartridges of different sizes to be interchanged using the same OMCA digital controller as a base
  • Cost-effective and versatile
Specifications
Capacity (Metric) 1000 mL
For Use With OMCA Digi-Mantle
Product Type Replacement Heating Cartridge