missing translation for 'onlineSavingsMsg'
Learn More

Repaglinide N-Oxide, TRC

Product Code. 30412035
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412035 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412035 Supplier Toronto Research Chemicals Supplier No. TRCR1445405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Repaglinide N-Oxide
CAS 121167-81-5
Molecular Formula C27 H36 N2 O5
Quantity 5 mg
Synonym Repaglinide N-Oxide, 2-Ethoxy-4-[2-[[3-methyl-1-[2-(1-oxido-1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]benzoic Acid, 2-Ethoxy-4-[2-[[3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]benzoic Acid N-Oxide
InChI Formula InChI=1S/C27H36N2O5/c1-4-34-25-17-20(12-13-22(25)27(31)32)18-26(30)28-23(16-19(2)3)21-10-6-7-11-24(21)29(33)14-8-5-9-15-29/h6-7,10-13,17,19,23H,4-5,8-9,14-16,18H2,1-3H3,(H,28,30)(H,31,32)/t23-/m0/s1
SMILES CCOc1cc(CC(=O)N[C@@H](CC(C)C)c2ccccc2[N+]3([O-])CCCCC3)ccc1C(=O)O
IUPAC Name 2-ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-oxidopiperidin-1-ium-1-yl)phenyl]butyl]amino]-2-oxoethyl]benzoic acid
Molecular Weight (g/mol) 468.585
Formula Weight 468.262
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.