Learn More
Trajan Scientific and Medical™ Removable Needle for Thermo Scientific RSH Autosampler
Replacement needles for use with Thermo Scientific RSH Autosampler
Brand:  Trajan Scientific and Medical™ 037871
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13199175
  • 44.00€ / Pack of 2
Estimated Shipment: 17-06-2024
Description
  • Replacement needle for use with Thermo Scientific RSH Autosampler
  • Removable needle
  • Cone Tip
Specifications
Specifications
23 G
6.3 mm
Removable Needle
2/Pk.
5.7 cm
Thermo RSH Autosampler
0.63 mm
57 mm
Cone tipped
10 μL
Cone