Learn More
Trajan Scientific and Medical™ Removable Needle for Thermo Scientific RSH Autosampler
Replacement needles for use with Thermo Scientific RSH Autosampler
43.20€ - 53.10€
Specifications
Product Type | Removable Needle |
---|---|
Needle Point Style | Cone |
For Use With (Equipment) | Thermo RSH Autosampler |
Product Code | Brand | Volume (Metric) | Needle Gauge | Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||
13119185
|
Trajan Scientific and Medical™
037875 |
10 μL | 26 G | 57 mm | 0.47 mm |
44.00€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13129185
|
Trajan Scientific and Medical™
037877 |
10 μL | 26 G | 85 mm | 0.47 mm |
53.10€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13139185
|
Trajan Scientific and Medical™
036875 |
5 μL | 26 G | 57 mm | 0.47 mm |
43.20€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13169175
|
Trajan Scientific and Medical™
036871 |
5 μL | 23 G | 57 mm | 0.63 mm |
44.00€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13179175
|
Trajan Scientific and Medical™
036873 |
5 μL | 23 G | 85 mm | 0.63 mm |
53.10€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13189175
|
Trajan Scientific and Medical™
036877 |
5 μL | 26 G | 85 mm | 0.47 mm |
53.10€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13199175
|
Trajan Scientific and Medical™
037871 |
10 μL | 23 G | 57 mm | 0.63 mm |
44.00€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
13109185
|
Trajan Scientific and Medical™
037873 |
10 μL | 23 G | 85 mm | 0.63 mm |
53.10€
Pack of 2 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
Description
- Replacement needle for use with Thermo Scientific RSH Autosampler
- Removable needle
- Cone Tip
Specifications
Removable Needle | |
Cone | |
Thermo RSH Autosampler |