Learn More
Trajan Scientific and Medical™ Removable Needle for Thermo Scientific RSH Autosampler
Replacement needles for use with Thermo Scientific RSH Autosampler
Brand:  Trajan Scientific and Medical™ 036873
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13179175
  • 53.10€ / Pack of 2
Estimated Shipment: 17-06-2024
Description
  • Replacement needle for use with Thermo Scientific RSH Autosampler
  • Removable needle
  • Cone Tip
Specifications
Specifications
23 G
0.63 mm
Removable Needle
5 μL
Thermo RSH Autosampler
0.63 mm
85 mm
2/Pk.
Cone