Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Product Code. 10785174 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
50 μL
Needle Gauge:
23 G
26 G
26s G
31 G
Length (Metric):
42 mm
50 mm
57 mm
75 mm
For Use With (Equipment):
AS2000 Autosampler
Agilent GC Instruments
CTC GC Instruments
Shimadzu GC Instruments
Thermo GCs
TriPlus RSH Autosampler
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Length (Metric) For Use With (Equipment) unitSize
10785174 10 μL 26 G 50 mm CTC GC Instruments Each
10702517 5 μL 31 G 75 mm Thermo GCs Each
10743867 10 μL 23 G 42 mm Agilent GC Instruments Each
10419054 10 μL 23 G 50 mm CTC GC Instruments Each
10724847 50 μL 23 G 50 mm AS2000 Autosampler Each
12896036 1 μL 23 G 57 mm TriPlus RSH Autosampler Each
15274563 5 μL 26s G 42 mm Shimadzu GC Instruments Each
15264573 10 μL 26s G 42 mm Shimadzu GC Instruments Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10785174 Supplier Thermo Scientific™ Supplier No. 365D1841
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Use Thermo Scientific™ Removable-Needle Syringes for accurate and reproducible sample introduction into GC instruments, with or without the needle.
Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Removable-Needle Syringes offer accurate and reproducible sample introduction, with or without the needle. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 26 G
Length (Metric) 50 mm
Product Type GC Autosampler Syringe, Removable Needle
Disposable Reusable
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) CTC GC Instruments
Replacement Needle 365RN362