Learn More
Thermo Scientific™ Removable-Needle Autosampler Syringes for Thermo Scientific™ Instruments
Product Code. 10702517 Shop All Thermo Scientific Products
Click to view available options
Volume (Metric):
1 μL
5 μL
10 μL
50 μL
Needle Gauge:
23 G
26 G
26s G
31 G
Length (Metric):
42 mm
50 mm
57 mm
75 mm
For Use With (Equipment):
AS2000 Autosampler
Agilent GC Instruments
CTC GC Instruments
Shimadzu GC Instruments
Thermo GCs
TriPlus RSH Autosampler
Unit Size:
Each
Product Code. 10702517
Brand: Thermo Scientific™ 36500510
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use Thermo Scientific™ Removable-Needle Syringes for accurate and reproducible sample introduction into GC instruments, with or without the needle.
Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Removable-Needle Syringes offer accurate and reproducible sample introduction, with or without the needle. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 31 G
Length (Metric) 75 mm
Product Type GC Autosampler Syringe, Removable Needle
Disposable Reusable
Volume (Metric) 5 μL
Length (Metric) Needle 75 mm
Needle Point Style Cone
For Use With (Equipment) Thermo GCs
Replacement Needle 36550045