Learn More
Thermo Scientific™ Remote Alarms for Ultra-Low Temperature Freezers
Increase sample security by using these remote alarms for ultra-low temperature freezers—designed for areas where an alarm may not be readily heard.
600.00€
Specifications
Description Remote Alarm with Audible and Visual Signal
Type Ultra-Low Freezer Remote Alarm
For Use With (Equipment) Ultra-Low Temperature Freezers
Monitoring Options Alarm
View More Specs
Products 1
Product Code Brand Voltage Price Quantity & Availability  
Product Code Brand Voltage Price Quantity & Availability  
13483918
View Documents Promotion Details
Thermo Scientific™
5612-1A
120 V This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
Increase sample security by using Thermo Scientific™ Remote Alarms for Ultra-Low Temperature Freezers — designed for areas where an alarm may not be readily heard.
Features:
  • Lightweight units attach to a wall or cabinet
  • Operate on line voltage
  • Switch to a battery backup if a power failure occurs
  • Can be connected easily to a temperature/power monitor and remote alarm contacts
  • Complete with audible alarm silence switch and alarm ringback features
  • Alarm can be temporarily defeated, but system rings back as a reminder to reset when normal conditions are restored
  • Audible and visual signal to indicate alarm condition on monitored equipment
Specifications
Specifications
Remote Alarm with Audible and Visual Signal
Ultra-Low Freezer Remote Alarm
Ultra-Low Temperature Freezers
Alarm