Learn More
Thermo Scientific™ Remote Alarms for Ultra-Low Temperature Freezers
Increase sample security by using these remote alarms for ultra-low temperature freezers—designed for areas where an alarm may not be readily heard.
600.00€
Specifications
Description | Remote Alarm with Audible and Visual Signal |
---|---|
Type | Ultra-Low Freezer Remote Alarm |
For Use With (Equipment) | Ultra-Low Temperature Freezers |
Monitoring Options | Alarm |
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
13483918
|
Thermo Scientific™
5612-1A |
120 V |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Increase sample security by using Thermo Scientific™ Remote Alarms for Ultra-Low Temperature Freezers — designed for areas where an alarm may not be readily heard.
Features:- Lightweight units attach to a wall or cabinet
- Operate on line voltage
- Switch to a battery backup if a power failure occurs
- Can be connected easily to a temperature/power monitor and remote alarm contacts
- Complete with audible alarm silence switch and alarm ringback features
- Alarm can be temporarily defeated, but system rings back as a reminder to reset when normal conditions are restored
- Audible and visual signal to indicate alarm condition on monitored equipment
Specifications
Remote Alarm with Audible and Visual Signal | |
Ultra-Low Freezer Remote Alarm | |
Ultra-Low Temperature Freezers | |
Alarm |