missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ YM Agar

Product Code. 15143898
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15143898 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15143898 Supplier Thermo Scientific™ Supplier No. R456032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Remel YM Agar is used for the cultivation of yeast, molds, and other aciduric microorganisms.

Remel YM Agar is used for the cultivation of yeast, molds, and other aciduric microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Description YM Agar
Form Powder
Product Type Agar
Quantity 500 g

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.