Learn More
Thermo Scientific™ Remel™ Sodium Carbonate
Product Code. 13277499
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13277499 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13277499 Supplier Thermo Scientific™ Supplier No. R21267
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Biochemically differentiate rapid and slow growing Mycobacterium spp. using Sodium Carbonate with the arylsulfatase test.
Biochemically differentiate rapid and slow growing Mycobacterium spp. using Thermo Scientific™ Remel™ Sodium Carbonate reagent with arylsulfatase test. Whitehead et al. described a liquid arylsulfatase test using phenolphthalein sulfate as the substrate, an incubation time of 2 weeks1 and could be used to differentiate virulent tubercle bacilli from nonvirulent strains. Wayne et al. reported that except Mycobacterium rhodochrous all mycobacteria produce the enzyme arylsulfatase2 and formulated liquid and solid arylsulfatase media3.
The arylsulfatase test measures the ability of certain mycobacteria to produce the enzyme arylsulfatase that attacks the substrate phenolphthalein sulfate, releasing free phenolphthalein. This leads to development of red color in the medium when sodium carbonate is added. Kubica and Rigdon proposed the use of 3-day arylsulfatase test to differentiate Mycobacterium fortuitum complex from saprophytic Mycobacterium spp.4Ready-to-use: Liquid reagentEasy to interpret: Pink-red color development / band on the surface is easy to interpret.This reagent contains 2N sodium carbonate (106 g/1000mL) and is supplied as 25 mL bottles.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Sodium Carbonate
Quantity 25 mL
Form Liquid