Learn More
Thermo Scientific™ Remel™ SAS™ Adeno Test
Description
Thermo Scientific™ Remel SAS Adeno Test is a cassette immunoassay used for qualitative detection of adenovirus antigens in eye swabs, nasal and pharyngeal secretions, and fecal material.
SA Scientific
Specifications
Specifications
| Description | SAS Adeno Test |
| Detectable Analytes | Adenovirus antigen |
| Quantity | 20 Tests/Kit |