Learn More
Thermo Scientific™ Remel™ SAS™ Adeno Test
Product Code. 11944062
Change view
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11944062 20 Tests/Kit 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11944062 Supplier Thermo Scientific™ Supplier No. R42067020
Cassette immunoassay used for qualitative detection of adenovirus antigens in eye swabs, nasal and pharyngeal secretions, and fecal material.
Thermo Scientific™ Remel SAS Adeno Test is a cassette immunoassay used for qualitative detection of adenovirus antigens in eye swabs, nasal and pharyngeal secretions, and fecal material.
SA Scientific
TRUSTED_SUSTAINABILITY
Specifications
Description SAS Adeno Test
Detectable Analytes Adenovirus antigen
Quantity 20 Tests/Kit